Skip to Content
Merck
All Photos(1)

Key Documents

111848

Sigma-Aldrich

trans-β-Methylstyrene

99%

Synonym(s):

trans-1-Phenyl-1-propene, trans-beta-Methylstyrene, trans-Propenylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$57.80
10 G
$350.00

$57.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$57.80
10 G
$350.00

About This Item

Linear Formula:
C6H5CH=CHCH3
CAS Number:
Molecular Weight:
118.18
Beilstein:
1361672
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$57.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

contains

20 ppm 3,5-di-tert-butylcatechol as inhibitor

refractive index

n20/D 1.550 (lit.)

bp

175 °C (lit.)

density

0.911 g/mL at 25 °C (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

C\C=C\c1ccccc1

InChI

1S/C9H10/c1-2-6-9-7-4-3-5-8-9/h2-8H,1H3/b6-2+

InChI key

QROGIFZRVHSFLM-QHHAFSJGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product exhibits potential exothermic hazards at higher temperatures[1].

Application

Trans-β-methylstyrene was used in the epoxidation reaction of an oxidant solution with a solution of dichloromethane, where nickel is used as a catalyst and benzyltributylammonium bromide as a phase-transfer agent[2].
Trans-β-methylstyrene was used in the preparation of exo-chromans by the oxa analogue of the Povarov reaction[3].

Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rivka R R Taylor et al.
The Journal of organic chemistry, 78(4), 1404-1420 (2012-12-13)
An oxa analogue of the well-known Povarov reaction has been developed for the synthesis of 3,4-dihydrobenzopyrans (chromans). The reaction involves the formal inverse electron demand [4 + 2] cycloaddition reaction of in situ-generated cationic aryl 2-oxadiene oxocarbenium ions with alkenes.
S Y Lin et al.
Journal of hazardous materials, 161(1), 330-335 (2008-05-20)
According to literature and our research, the styrene polymerization mechanism is identified by alpha-methylstyrene (AMS). This study investigated the basic exothermic behavior of styrene and its major derivatives, AMS, and trans-beta-methylstyrene (TBMS), by two calorimeters, differential scanning calorimetry (DSC) and
(Salen)nickel-Catalysed Epoxidations in the Homogeneous and Heterogeneous Phase: The Implications of Oxygen on the Efficiency and Product Selectivity.
Ferreira R, et al.
European Journal of Inorganic Chemistry, 2005(21), 4272-4279 (2005)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service