Skip to Content
Merck
All Photos(1)

Key Documents

414077

Sigma-Aldrich

Poly(vinyl alcohol-co-ethylene)

ethylene 27 mol %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)x[CH2CH(OH)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

melt index

3.9 g/10 min (210°C)

composition

ethylene, 27 mol %

hardness

104 (Rockwell M, ASTM D 785)

impurities

<0.7% polymerized vinyl acetate

transition temp

Tm 191 °C (DSC peak)
Tg 72 °C

solubility

DMSO, water/n-propanol (1:1), and phenol: soluble

density

1.2 g/mL at 25 °C

SMILES string

C=C.OC=C

InChI

1S/C2H4O.C2H4/c1-2-3;1-2/h2-3H,1H2;1-2H2

InChI key

UFRKOOWSQGXVKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl alcohol-co-ethylene) (PVA-co-PE) is a hydrophilic non-toxic polymer with good film forming ability. Its other properties include biocompatibility, high tensile strength and low fouling potential.[1]

Application

PVA-co-PE can be used in the formation of nanofibrous membranes for the development of high performance adsorption materials.[2] It can also be used to form membranes for potential applications such as lithium ion batteries, biosensors and microfluidic devices.[3][4]

Other Notes

Maximum working temperature 240°C
See 414093 as an alternate for this product.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Light-induced switching of the wettability of novel asymmetrical poly (vinyl alcohol)-co-ethylene membranes blended with azobenzene polymers
Tylkowski B, et al.
Langmuir, 26(18), 14821-14829 (2010)
Cibacron Blue F3GA functionalized poly (vinyl alcohol-co-ethylene)(PVA-co-PE) nanofibrous membranes as high efficient affinity adsorption materials
Zhu J, et al.
Journal of Membrane Science , 385, 269-276 (2011)
A novel hierarchically structured and highly hydrophilic poly (vinyl alcohol-co-ethylene)/poly (ethylene terephthalate) nanoporous membrane for lithium-ion battery separator
Xia M, et al.
Journal of Power Sources, 266, 29-35 (2014)
High performance filtration nanofibrous membranes based on hydrophilic poly (vinyl alcohol-co-ethylene) copolymer
Li M, et al.
Desalination, 329, 50-56 (2013)
C-Y Huang et al.
IET nanobiotechnology, 5(4), 126-131 (2011-12-14)
Neopterin is a catabolic product of guanosine triphosphate, a purine nucleotide. Measuring neopterin concentrations in biological fluids such as urine provides information about cellular immune activation in humans under control of T helper cells. A high neopterin concentration in bodily

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service