Skip to Content
Merck
All Photos(2)

Key Documents

191108

Sigma-Aldrich

Poly(styrene-co-allyl alcohol)

average Mn ~1,600, crystalline, allyl alcohol 33 mol %

Synonym(s):

Allyl alcohol-styrene copolymer, Poly(styrene-allyl alcohol), Styrene-allyl alcohol copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$271.00

$271.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$271.00

About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2CH(CH2OH)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$271.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

2 (vs air)

vapor pressure

23.8 mmHg ( 25 °C)

form

crystalline

mol wt

average Mn ~1,600

composition

allyl alcohol, 33 mol %

expl. lim.

18 %

density

1.083 g/mL at 25 °C

SMILES string

OCC=C.C=Cc1ccccc1

InChI

1S/C8H8.C3H6O/c1-2-8-6-4-3-5-7-8;1-2-3-4/h2-7H,1H2;2,4H,1,3H2

InChI key

WXNYILVTTOXAFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service