Skip to Content
Merck
All Photos(3)

Key Documents

413275

Sigma-Aldrich

2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol

96%

Synonym(s):

(3,5-di-tert -Butyl-4-hydroxybenzyl)dimethylamine, 2,6-Di-tert -butyl-α-(dimethylamino)-4-cresol, 4-[(Dimethylamino)methyl]-2,6-bis(1,1-dimethylethyl)phenol, 4-[(Dimethylamino)methyl]-2,6-di-tert -butylphenol

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$114.00
1 KG
$300.00

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$114.00
1 KG
$300.00

About This Item

Linear Formula:
(CH3)2NCH2C6H2[C(CH3)3]2OH
CAS Number:
Molecular Weight:
263.42
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

bp

172 °C/30 mmHg (lit.)

mp

90-95 °C

SMILES string

CN(C)Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C17H29NO/c1-16(2,3)13-9-12(11-18(7)8)10-14(15(13)19)17(4,5)6/h9-10,19H,11H2,1-8H3

InChI key

VMZVBRIIHDRYGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Di-tert-butyl-4-(dimethylaminomethyl)phenol is a hindered phenolic compound that is used as an antioxidant to stabilize the lubricant oils.[1][2]

Application

Antioxidant

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth and characterization of 2, 6-Di-tert-butyl-4-(dimethylaminomethyl) phenol single crystal by the vertical Bridgman method.
SolankiSSB, et al.
Journal of Crystal Growth, 411(24), 19-23 (2015)
Electrospray ionization and atmospheric pressure matrix-assisted laser desorption/ionization mass spectrometry of antioxidants applied in lubricants.
Kassler A, et al.
Rapid Communications in Mass Spectrometry, 23(24), 3917-3927 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service