Skip to Content
Merck
All Photos(1)

Key Documents

412015

Sigma-Aldrich

3-Bromo-4-methoxybenzaldehyde

98%

Synonym(s):

3-Bromo-p-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
215.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

51-54 °C (lit.)

SMILES string

COc1ccc(C=O)cc1Br

InChI

1S/C8H7BrO2/c1-11-8-3-2-6(5-10)4-7(8)9/h2-5H,1H3

InChI key

QMPNFQLVIGPNEI-UHFFFAOYSA-N

General description

3-Bromo-4-methoxybenzaldehyde is formed by the solvent-free bromination of 4-methoxybenzaldehyde using 1,3-di-n-butylimidazolium tribromide, as a brominating reagent.

Application

3-Bromo-4-methoxybenzaldehyde may be used in the following studies:
  • Asymmetric synthesis of a novel β-hydroxy-α-amino acid derivative, via Mukaiyama aldol reaction.
  • Synthesis of 2-(3-bromo-4-methoxyphenyl)-5-fluorobenzothiazole.
  • Preparation of 5-[(Z)-2-(3-bromo-4-methoxyphenyl)vinyl]-1,2-3-trimethoxybenzene.
  • Total synthesis of engelhardione.
  • Starting reagent for the synthesis of (2E)-3-(3-bromo-4-methoxyphenyl)-1-(4-methylphenyl)prop-2-en-1-one.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

22287813
10th Internatl. Conf. on Organic Synthesis, Bangalore, India, December 1994 null
(2E)-3-(3-Bromo-4-methoxyphenyl)-1-(4-methylphenyl)prop-2-en-1-one.
Dutkiewicz g, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(4), 1024-1024 (2011)
Li Shen et al.
Tetrahedron letters, 52(35), 4570-4574 (2011-09-20)
The total synthesis of the macrocyclic natural product engelhardione is reported. This effort led to the structural revision of the published structure of engelhardione to that of pterocarine. The revision reflects the change of the substitution pattern of one phenyl
Aromatic bromination of aldehydes and ketones using 1, 3-di-n-butylimidazolium tribromide [BBIm] Br3 ionic liquids under solvent-free conditions.
Borikar SP and Daniel T.
Journal of the Iranian Chemical Society, 8(2), 531-536 (2011)
Yali Kong et al.
Chemistry & biology, 12(9), 1007-1014 (2005-09-27)
Targeting the microtubule system represents an attractive strategy for the development of anticancer agents. In this study, we report a class of combretastatin A-4 (CA-4) analogs derivatized with a boronic acid moiety replacing the hydroxyl group on the C-ring of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service