Skip to Content
Merck
All Photos(1)

Key Documents

411531

Sigma-Aldrich

(Bromomethylene)dimethyliminium bromide

95%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$494.00

$494.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$494.00

About This Item

Linear Formula:
BrCH=N(CH3)2(Br)
CAS Number:
Molecular Weight:
216.90
Beilstein:
4155730
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$494.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

solid

mp

148-153 °C (lit.)

functional group

amine
bromo

SMILES string

[Br-].C\[N+](C)=C\Br

InChI

1S/C3H7BrN.BrH/c1-5(2)3-4;/h3H,1-2H3;1H/q+1;/p-1

InChI key

XIXYNENNFUBWFK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

(Bromomethylene)dimethyliminium bromide participates in one-carbon homologation of aldehydes to carboxylic acid derivatives.[1]

Application

(Bromomethylene)dimethyliminium bromide (bromodi- methylformiminium bromide) may be employed for the synthesis of 6-phenyl-6-deoxy-2,3-di-O-methylcellulose.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a one-pot method for the homologation of aldehydes to carboxylic acids.
McNulty J and Das P.
Tetrahedron, 65(37), 7794-7800 (2009)
Synthesis and NMR characterization of 6-Phenyl-6-deoxy-2, 3-di-O-methylcellulose.
Winsor DL, et al.
Polymers For Advanced Technologies, 13(6), 413-427 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service