410187
Poly(propylene glycol) methyl ether acrylate
average Mn ~260, tripropylene glycol acrylate ~30 %
Synonym(s):
Methoxy polypropylene glycol monoacrylate
About This Item
Recommended Products
mol wt
average Mn ~260
contains
250 ppm MEHQ as inhibitor
composition
tripropylene glycol acrylate, ~30%
refractive index
n20/D 1.433 (lit.)
density
0.987 g/mL at 25 °C (lit.)
SMILES string
COCCCOCCCOCCCOC(=O)C=C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service