Skip to Content
Merck
All Photos(1)

Key Documents

40980

Sigma-Aldrich

3,7-Dimethyl-1-octene

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$107.00

$107.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$107.00

About This Item

Linear Formula:
(CH3)2CHCH2CH2CH2CH(CH3)CH=CH2
CAS Number:
Molecular Weight:
140.27
Beilstein:
1719870
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$107.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

refractive index

n20/D 1.417

bp

154-156 °C (lit.)

density

0.733 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCCC(C)C=C

InChI

1S/C10H20/c1-5-10(4)8-6-7-9(2)3/h5,9-10H,1,6-8H2,2-4H3

InChI key

KSXTZYRIJKDCEA-UHFFFAOYSA-N

General description

(R)-3,7-dimethyl-1-octene is an optically active alkyl-α-olefin. Its copolymerization with styrene, 1-vinylnaphthalene or 2-methylstyrene by Ziegler-Natta catalysts has been reported.[1] Preparation of copolymers of racemic and optically active 3,7-dimethyl-1-octene with ethylene in the presence of Ziegler-Natta isospecific catalysts have been studied.[2] Polymerization of (R,S)-3,7-dimethyl-1-octene in the presence of heterogeneous or homogeneous isotactic specific catalysts involves highly stereospecific insertion of (R,S)-3,7-dimethyl-1-octene into the metal-CH3 bond.[3]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

13C-enriched end groups of poly (3, 7-dimethyl-1-octene) prepared in the presence of isotactic specific catalysts.
Longo P, et al.
Macromolecular Rapid Communications, 18(6), 491-495 (1997)
Stereoselective copolymerization of racemic a-olefins with ethylene by isospecific Ziegler-Natta catalyst.
Ciardelli F, et al.
Marine Chemistry, 175(3), 923-933 (1974)
Optically active hydrocarbon polymers with aromatic side chains, 7. Synthesis and properties of copolymers of vinyl aromatic monomers with chiral α-olefins.
Carlini C and Chiellini E.
Marine Chemistry, 176(3), 519-535 (1975)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service