Skip to Content
Merck
All Photos(1)

Key Documents

409421

Sigma-Aldrich

3-Allyloxy-2-hydroxy-1-propanesulfonic acid sodium salt solution

40 wt. % in H2O

Synonym(s):

1-Propanesulfonic acid, 2-hydroxy-3-(2-propenyloxy)-, monosodium salt, Sodium 3-allyloxy-2-hydroxy-1-propanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$90.40

$90.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$90.40

About This Item

Linear Formula:
H2C=CHCH2OCH2CH(OH)CH2SO3Na
CAS Number:
Molecular Weight:
218.20
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$90.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

concentration

40 wt. % in H2O

refractive index

n20/D 1.394

density

1.17 g/mL at 25 °C

SMILES string

[Na+].OC(COCC=C)CS([O-])(=O)=O

InChI

1S/C6H12O5S.Na/c1-2-3-11-4-6(7)5-12(8,9)10;/h2,6-7H,1,3-5H2,(H,8,9,10);/q;+1/p-1

InChI key

CJGJYOBXQLCLRG-UHFFFAOYSA-M

General description

3-Allyloxy-2-hydroxy-1-propane sulfonic acid sodium salt solution is a water-soluble compound characterized by the presence of an allyloxy group and a sulfonic acid functional group. This compound is known for its excellent solubility in aqueous environments, making it suitable for various applications in chemical and pharmaceutical formulations.

Application

3-Allyloxy-2-hydroxy-1-propanesulfonic acidsodium salt (AHPS) can be used as:
  • A monomer in the synthesis of a hydrogel through free radical-induced polymerization with acrylamide. The primary use of AHPS is to introduce sulfonate groups into the hydrogel structure, enhancing its ability to adsorb methylene blue dye.
  • A surface modification and functionalization agent in the development of polymeric carbon nitride nanotubes which find various applications, including biosensing, drug delivery, and environmental monitoring.
  • A key component in the formulation of the nanoparticles. Its properties as a surfactant may enhance the stability anddispersion of the nanoparticles in aqueous environments, facilitating their application in controlled release systems.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service