Skip to Content
Merck
All Photos(2)

Key Documents

407852

Sigma-Aldrich

tert-Butyldimethylsilyl cyanide

97%

Synonym(s):

tert-Butyl-cyano-dimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2CN
CAS Number:
Molecular Weight:
141.29
Beilstein:
2234709
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

76-79 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)C#N

InChI

1S/C7H15NSi/c1-7(2,3)9(4,5)6-8/h1-5H3

InChI key

CWAKIXKDPQTVTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyldimethylsilyl cyanide (TBDMSCN) is a bulkier trialkylsilylcyanide.[1] It participates in the cyanosilylation of enantiopure 4-oxoazetidine-2-carbaldehydes.[2] Addition of TBDMSCN to sterically hindered ketones in the presence of Lewis acid or base catalyst has been studied. ZnI2-catalyzed addition of TBDSCN to 2,2-dimethylcyclohexanone, 2,2,6-trimethylcyclohexanone and 2,2,6,6-tetramethylcyclohexanone affords protected cyanohydrins.[3]

Application

tert-Butyldimethylsilyl cyanide may be used as reagent for the formation of β-isonitrile alcohols via epoxides.[4]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 51, 5010-5010 (1986)
P(RNCH2CH2)N: efficient catalysts for the cyanosilylation of aldehydes and ketones.
Fetterly BM and Verkade JG.
Tetrahedron Letters, 46(46), 8061-8066 (2005)
Addition of tert-butyldimethyl-or tert-butyldiphenylsilyl cyanide to hindered ketones.
Golinski M, et al.
The Journal of Organic Chemistry, 58(1), 159-164 (1993)
Benito Alcaide et al.
The Journal of organic chemistry, 72(21), 7980-7991 (2007-09-18)
The cyanosilylation of enantiopure 4-oxoazetidine-2-carbaldehydes with tert-butyldimethylsilyl cyanide was promoted by either molecular sieves or catalytic amount of sodium carbonate to give O-silylated beta-lactam cyanohydrins with good yield and diastereoselectivity. In contrast, Lewis acids did not effectively promote the cyanosilylation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service