Skip to Content
Merck
All Photos(2)

Key Documents

407429

Sigma-Aldrich

Methyl 3-methoxypropionate

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$84.50
1 L
$189.00

$84.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$84.50
1 L
$189.00

About This Item

Linear Formula:
CH3OCH2CH2COOCH3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1744829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

liquid

refractive index

n20/D 1.402 (lit.)

bp

142-143 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

COCCC(=O)OC

InChI

1S/C5H10O3/c1-7-4-3-5(6)8-2/h3-4H2,1-2H3

InChI key

BDJSOPWXYLFTNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 3-methoxypropionate is an ester.

Application

Methyl 3-methoxypropionate may be employed as acylation reagent for the lipase-catalyzed N-acylation of 1-phenylethanamine.[1] It may be used in the synthesis of poly(2-hydroxylethyl 5-norbornene-2-carboxylate /t-butyl 5-norbornene-2-carboxylate /5-norbornene-2-carboxylic acid /maleic anhydride) resists. Lithographic performance of these resists was studied using ArF stepper.[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Alicyclic Polymers for 193nm Single Layer Resist Materials
Jung J-C, et al.
J. Photopolym. Sci. Technol., 11(3), 481-488 (1998)
Maria Cammenberg et al.
Chembiochem : a European journal of chemical biology, 7(11), 1745-1749 (2006-09-23)
One of the commercial methods for preparing enantiopure amines is lipase-catalyzed kinetic resolution, although lipases catalyze aminolysis with only low activity. Interestingly, in 1997 Balkenhohl et al. used ethyl methoxyacetate instead of ethyl butyrate as an acylation reagent for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service