Skip to Content
Merck
All Photos(4)

Documents

270121

Sigma-Aldrich

Methyl (S)-(+)-3-hydroxy-2-methylpropionate

97%

Synonym(s):

(+)-Methyl L-β-hydroxyisobutyrate, (S)-(+)-3-Hydroxy-2-methylpropionic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(CH3)CO2CH3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1720830
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

optical activity

[α]19/D +26°, c = 4 in methanol

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.425 (lit.)

bp

74 °C/10 mmHg (lit.)

density

1.071 g/mL at 25 °C (lit.)

SMILES string

COC(=O)[C@@H](C)CO

InChI

1S/C5H10O3/c1-4(3-6)5(7)8-2/h4,6H,3H2,1-2H3/t4-/m0/s1

InChI key

ATCCIZURPPEVIZ-BYPYZUCNSA-N

Application

Methyl (S)-(+)-3-hydroxy-2-methylpropionate may be used as a starting material in the total synthesis of (+)-discodermolide, a potent antimitotic agent.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Total synthesis of the antimicrotubule agent (+)-discodermolide using boron-mediated aldol reactions of chiral ketones.
Paterson I, et al.
Angewandte Chemie (International Edition in English), 39(2), 377-380 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service