Skip to Content
Merck
All Photos(2)

Key Documents

407216

Sigma-Aldrich

Triarylsulfonium hexafluorophosphate salts, mixed

50% in propylene carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$72.50
100 ML
$242.00

$72.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$72.50
100 ML
$242.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$72.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

concentration

50% in propylene carbonate

refractive index

n20/D 1.5 (lit.)

density

1.32 g/mL at 25 °C (lit.)

SMILES string

F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.S(c1ccccc1)c2ccc(cc2)[S+](c3ccccc3)c4ccccc4.S(c5ccc(cc5)[S+](c6ccccc6)c7ccccc7)c8ccc(cc8)[S+](c9ccccc9)c%10ccccc%10

InChI

1S/C36H28S3.C24H19S2.3F6P/c1-5-13-31(14-6-1)38(32-15-7-2-8-16-32)35-25-21-29(22-26-35)37-30-23-27-36(28-24-30)39(33-17-9-3-10-18-33)34-19-11-4-12-20-34;1-4-10-20(11-5-1)25-21-16-18-24(19-17-21)26(22-12-6-2-7-13-22)23-14-8-3-9-15-23;3*1-7(2,3,4,5)6/h1-28H;1-19H;;;/q+2;+1;3*-1

InChI key

BXEZIWXFNSCYKD-UHFFFAOYSA-N

General description

Mixed triarylsulfonium hexafluorophosphate salts are cationic photoinitiators which are used as ultraviolet curing agents that have bifunctional cross-linking monomers.[1]

Application

It can be mixed with propylene glycol monomethyl ether acetate (PGMA) that may be blended with bisphenol F resin which can be used in the fabrication of UV curable nano-imprinted epoxy resist.[2][3] It can also be used as a photo-initiator for photo-polymerization of epoxy/acrylate composites which find applications in UV protective coating on electronic devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel visible photoinitiators systems for free-radical/cationic hybrid photopolymerization.
Podsiadly R.
Dyes and Pigments, 91(3), 422-426 (2011)
Nanoimprinting ultrasmall and high-aspect-ratio structures by using rubber-toughened UV cured epoxy resist.
Shin YJ, et al.
Nanotechnology, 24(25), 255302-255302 (2013)
Kinetics study of the UV-initiated cationic polymerization of cycloaliphatic diepoxide resins.
Voytekunas VY, et al.
European Polymer Journal, 44(11), 3640-3649 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service