Skip to Content
Merck
All Photos(2)

Key Documents

404659

Sigma-Aldrich

Tin(IV) bis(acetylacetonate) dichloride

98%

Synonym(s):

Bis(acetylacetonato)dichlorotin, Dichlorotin bis(acetylacetonate)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$153.00

$153.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$153.00

About This Item

Linear Formula:
[CH3COCH=C(O-)CH3]2SnCl2
CAS Number:
Molecular Weight:
387.83
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$153.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

flakes
powder or crystals

reaction suitability

core: tin

mp

204-206 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Sn](Cl)(Cl)O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.2ClH.Sn/c2*1-4(6)3-5(2)7;;;/h2*3,6H,1-2H3;2*1H;/q;;;;+4/p-4/b2*4-3-;;;

InChI key

DTMARTPDQMQTRX-VGKOASNMSA-J

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service