Skip to Content
Merck
All Photos(2)

Key Documents

339059

Sigma-Aldrich

Barium acetylacetonate hydrate

Synonym(s):

Ba(acac)2, Barium 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$254.00

$254.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
50 G
$254.00

About This Item

Linear Formula:
Ba(C5H7O2)2 · xH2O
Molecular Weight:
335.54 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$254.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder or crystals (or Flakes or Beads)

composition

Ba, ≥30.0% gravimetric

reaction suitability

core: barium

SMILES string

O.CC(=O)\C=C(\C)O[Ba]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Ba.H2O/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;1H2/q;;+2;/p-2/b2*4-3-;;

InChI key

HYRVTHFPMWHAEG-SUKNRPLKSA-L

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service