Skip to Content
Merck
All Photos(1)

Key Documents

403458

Sigma-Aldrich

N-p-Tosylglycine

97%

Synonym(s):

N-(p-Toluenesulfonyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NHCH2CO2H
CAS Number:
Molecular Weight:
229.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

147-149 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cc1ccc(cc1)S(=O)(=O)NCC(O)=O

InChI

1S/C9H11NO4S/c1-7-2-4-8(5-3-7)15(13,14)10-6-9(11)12/h2-5,10H,6H2,1H3,(H,11,12)

InChI key

VDKFCCZUCXYILI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sener et al.
Biochemical pharmacology, 34(14), 2495-2499 (1985-07-15)
N-p-tosylglycine, which inhibits transglutaminase activity in islet homogenates, was found to cause a rapid and sustained facilitation of insulin release evoked by D-glucose, L-leucine or the association of Ba2+ and theophylline in intact islets. Such a facilitating action could not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service