Skip to Content
Merck
All Photos(2)

Key Documents

403318

Sigma-Aldrich

Indium(III) selenide

Synonym(s):

Indium selenide, Indium sesquiselenide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$426.00

$426.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$426.00

About This Item

Linear Formula:
In2Se3
CAS Number:
Molecular Weight:
466.52
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

$426.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

density

5.67 g/mL at 25 °C (lit.)

SMILES string

[Se]=[In][Se][In]=[Se]

InChI

1S/2In.3Se

InChI key

VSXOUQNLLPQTQK-UHFFFAOYSA-N

Related Categories

General description

Indium selenide (In2Se3) exists in 4 crystal structures (α,β,δ,γ).The metallic α-phase of indium selenide converts to intrinsic conduction β-phase under change in temperature. In2Se3 can be prepared by stoichiometric combination of the elements at 1000 to 1100oC temperatures. Influence of phase transformations on its optical and electrical properties has been reported.[1]

Packaging

Packaged in glass bottles

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth of single-phase In2Se3 by using metal organic chemical vapor deposition with dual-source precursors.
Chang KJ, et al.
Applied Physics Letters, 89(18), 2118-2118 (2006)
Some electrical and optical properties of In2Se3
Bidjin DS, et al.
Physica Status Solidi (A): Applied Research, 6(1), 295-299 (1971)

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service