Skip to Content
Merck
All Photos(1)

Documents

402737

Sigma-Aldrich

Magnesium phthalocyanine

Dye content 90 %

Synonym(s):

Phthalocyanine magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16MgN8
CAS Number:
Molecular Weight:
536.83
Beilstein:
3584903
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 90%

λmax

668 nm

SMILES string

[Mg]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Mg/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

NIQREIWCURIAMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jane P Bearinger et al.
Langmuir : the ACS journal of surfaces and colloids, 25(2), 1238-1244 (2008-12-31)
Photocatalytic lithography (PCL) is an inexpensive, fast, and robust method of oxidizing surface chemical moieties to produce patterned substrates. This technique has utility in basic biological research as well as various biochip applications. We report on porphyrin-based PCL for patterning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service