Skip to Content
Merck
All Photos(3)

Key Documents

252980

Sigma-Aldrich

Copper(II) phthalocyanine

β-form, Dye content 90 %

Synonym(s):

CuPc, Phthalocyanine blue, Pigment Blue 15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16CuN8
CAS Number:
Molecular Weight:
576.07
Colour Index Number:
74160
Beilstein:
4121848
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

composition

Dye content, 90%

λmax

678 nm

OLED Device Performance

ITO/CuPc/NPD/Alq3/C60/Mg:Ag

  • Color: green
  • Max. Luminance: 17200 Cd/m2

ITO/CuPc/NPD/Alq3/LiF/Al
  • Color: red
  • Max. Luminance: 2000 Cd/m2
  • Turn-On Voltage: 7.5 V

ITO/CuPc/NPD/CBP:FIrpic (6%)/BAlq3/LiF/Al
  • Color: blue
  • Max. EQE: 4.5 %

OPV Device Performance

ITO/CuPc/PTCDA/In

  • Short-circuit current density (Jsc): 53.1 mA/cm2
  • Open-circuit voltage (Voc): 0.57 V
  • Fill Factor (FF): 0.61
  • Power Conversion Efficiency (PCE): 4.2 %

ITO/PEDOT:PSS/CuPc/C60/BCP/Al
  • Short-circuit current density (Jsc): 18.8 mA/cm2
  • Open-circuit voltage (Voc): 0.58 V
  • Fill Factor (FF): 0.52
  • Power Conversion Efficiency (PCE): 3.6 %

SMILES string

c1ccc2c(c1)C3=NC4=[N@@H]5C(=Nc6n7c(N=C8c9ccccc9C%10=[N@@H]8[Cu]57N3C2=N%10)c%11ccccc6%11)c%12ccccc4%12

InChI

1S/C32H16N8.Cu/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

XCJYREBRNVKWGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary J Comeau et al.
ACS sensors, 4(10), 2706-2715 (2019-08-28)
Quality control is imperative for Cannabis since the primary cannabinoids, Δ9-tetrahydrocannabinol (THC) and cannabidiol (CBD), elicit very different pharmacological effects. THC/CBD ratios are currently determined by techniques not readily accessible by consumers or dispensaries and which are impractical for use
Alexander J Peltekoff et al.
Polymers, 12(6) (2020-06-04)
A library of statistically random pentafluorostyrene (PFS) and methyl methacrylate (MMA) copolymers with narrow molecular weight distributions was produced, using nitroxide mediated polymerization (NMP) to study the effect of polymer composition on the performance of bottom-gate top-contact organic thin-film transistors
Jaron G Van Dijken et al.
Molecules (Basel, Switzerland), 17(9), 10119-10130 (2012-08-28)
We investigate the evolution of copper phthalocyanine thin films as they are etched with argon plasma. Significant morphological changes occur as a result of the ion bombardment; a planar surface quickly becomes an array of nanopillars which are less than
Richard Murdey et al.
The Journal of chemical physics, 134(23), 234702-234702 (2011-06-28)
The current flowing through a thin film of copper phthalocyanine vacuum deposited on a single crystal sapphire [0001] surface was measured during film growth from 0 to 93 nm. The results, expressed as conductance vs. nominal film thickness, indicate three
Kai Xiao et al.
Journal of the American Chemical Society, 135(9), 3680-3687 (2013-02-02)
The epitaxial growth and preferred molecular orientation of copper phthalocyanine (CuPc) molecules on graphene has been systematically investigated and compared with growth on Si substrates, demonstrating the role of surface-mediated interactions in determining molecular orientation. X-ray scattering and diffraction, scanning

Articles

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service