Skip to Content
Merck
All Photos(1)

Key Documents

402478

Sigma-Aldrich

2,2,2,4′-Tetrafluoroacetophenone

99%

Synonym(s):

α,α,α,4-Tetrafluoroacetophenone

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$183.00

$183.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$183.00

About This Item

Linear Formula:
FC6H4COCF3
CAS Number:
Molecular Weight:
192.11
Beilstein:
1872824
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$183.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

66-67 °C/34 mmHg (lit.)

mp

24-27 °C (lit.)

density

1.37 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

Fc1ccc(cc1)C(=O)C(F)(F)F

InChI

1S/C8H4F4O/c9-6-3-1-5(2-4-6)7(13)8(10,11)12/h1-4H

InChI key

LUKLMXJAEKXROG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,2,4′-Tetrafluoroacetophenone (α,α,α,4-Tetrafluoroacetophenone, 2,2,2-trifluoro-1-(4-fluorophenyl)ethanone) is a fluorinated building block. Its enthalpy of vaporization at boiling point has been reported.[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proline Catalyzed Asymmetric Aldol Reaction between Methyl Ketones and 1-Aryl-2, 2, 2-trifluoroethanones.
Li-Hua Q, et al
Chin. J. Chem., 23(5), 584-588 (2005)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 548-548 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service