Skip to Content
Merck
All Photos(1)

Documents

40241

Sigma-Aldrich

(S)-(−)-N,N-Dimethyl-1-ferrocenylethylamine

≥98.0% (T)

Synonym(s):

(S)-(−)-[1-(Dimethylamino)ethyl]ferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19FeN
CAS Number:
Molecular Weight:
257.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

optical activity

[α]20/D −14±1°, c = 1.5% in ethanol

color

dark brown

bp

120-121 °C/0.7 mmHg (lit.)

density

1.222 g/mL at 20 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][CH]2)N(C)C

InChI

1S/C9H14N.C5H5.Fe/c1-8(10(2)3)9-6-4-5-7-9;1-2-4-5-3-1;/h4-8H,1-3H3;1-5H;/t8-;;/m0../s1

InChI key

ISOLNZQTVMCEOI-JZGIKJSDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(−)-N,N-Dimethyl-1-ferrocenylethylamine can be used:
  • As a starting material in the synthesis of chiral diferrocenylphosphine-diamines applicable in catalysis.
  • As a model chiral probe in the study of enantiodiscrimination ability of chiral ionic liquids using cyclic voltammetry.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Diastereoselective Synthesis of Chiral Diferrocenylphosphine-diamines: X-ray Crystal Structure of [(η5-C5H5) Fe
Zhou Z and Zhang Y
Synthetic Communications, 35(18), 2401-2408 (2005)
A family of chiral ionic liquids from the natural pool: Relationships between structure and functional properties and electrochemical enantiodiscrimination tests
Longhi M, et al.
Electrochimica Acta, 298(18), 194-209 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service