Skip to Content
Merck
All Photos(1)

Key Documents

372625

Sigma-Aldrich

1,1′-Ferrocenedimethanol

97%

Synonym(s):

1,1′-Ferrocenylbis(methanol), Cyclopentadienemethanol

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$358.00

$358.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$358.00

About This Item

Empirical Formula (Hill Notation):
C12H14FeO2
CAS Number:
Molecular Weight:
246.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$358.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

reaction suitability

core: iron
reagent type: catalyst

mp

106-108 °C (lit.)

SMILES string

[Fe].OC[C]1[CH][CH][CH][CH]1.OC[C]2[CH][CH][CH][CH]2

InChI

1S/2C6H7O.Fe/c2*7-5-6-3-1-2-4-6;/h2*1-4,7H,5H2;

InChI key

ZFFYTNSKVXIYIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haipeng Xiang et al.
Nanoscale, 10(28), 13572-13580 (2018-07-06)
A sandwich-type electrochemical immunosensor for sensitive detection of a tumor marker, α-fetoprotein (AFP), was fabricated by employing polydopamine-detection antibody nanoparticles (PDANPs-Ab2) as selective redox cycling-based signal amplifiers on an electrodeposited nano-gold electrode. In this research, PDANPs prepared through oxidative polymerization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service