Skip to Content
Merck
All Photos(1)

Key Documents

40199

Sigma-Aldrich

Methyl tetrahydro-2H-pyran-4-carboxylate

produced by BASF, ≥98.0% (GC)

Synonym(s):

THPE

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$204.00

About This Item

Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
Beilstein:
114974
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

produced by BASF

Assay

≥98.0% (GC)

form

liquid

density

1.080 g/mL at 20 °C (lit.)

functional group

ester
ether

SMILES string

COC(=O)C1CCOCC1

InChI

1S/C7H12O3/c1-9-7(8)6-2-4-10-5-3-6/h6H,2-5H2,1H3

InChI key

CNCMVGXVKBJYNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl tetrahydro-2H-pyran-4-carboxylate may be used in the following syntheses:
  • cycloalkylamide derivatives[1]
  • tetrahydro-2-furoic chloride
  • tetrahydro-2H-pyran-4-carboxylic chloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-Hae Kim et al.
Journal of medicinal chemistry, 54(6), 1752-1761 (2011-02-23)
Structure-activity relationships of cycloalkylamide compounds as inhibitors of human sEH were investigated. When the left side of amide function was modified by a variety of cycloalkanes, at least a C6 like cyclohexane was necessary to yield reasonable inhibition potency on
Han Xiao et al.
Environmental pollution (Barking, Essex : 1987), 260, 113962-113962 (2020-02-01)
Tris (4-hydroxyphenyl)ethane (THPE), a trisphenol compound widely used as a branching agent and raw material in plastics, adhesives, and coatings is rarely regarded with concern. However, inspection of in vitro data suggests that THPE is an antagonist of estrogen receptors (ERs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service