Skip to Content
Merck
Methyl tetrahydro-2H-pyran-4-carboxylate produced by BASF, ≥98.0% (GC)

Methyl tetrahydro-2H-pyran-4-carboxylate

Methyl tetrahydro-2H-pyran-4-carboxylate produced by BASF, ≥98.0% (GC)
Synonym(s):
THPE
Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing