Skip to Content
Merck
All Photos(2)

Key Documents

401293

Sigma-Aldrich

Rubidium hydroxide hydrate

Synonym(s):

Rubidium hydroxide, Rubidium(1+) hydroxide hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$137.00
25 G
$580.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$137.00
25 G
$580.00

About This Item

Linear Formula:
RbOH · xH2O
CAS Number:
Molecular Weight:
102.48 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

$137.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

crystals and lumps

Quality Level

composition

Degree of hydration, ~1

concentration

≤15.4% (% OH, titration by HCl)

impurities

≤15% Rb2CO3

SMILES string

O.[OH-].[Rb+]

InChI

1S/2H2O.Rb/h2*1H2;/q;;+1/p-1

InChI key

CHAWVFWLJTUJNI-UHFFFAOYSA-M

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liangying Li et al.
ACS applied materials & interfaces, 11(2), 2543-2550 (2018-12-20)
The demand for CO2/C2H2 separation, especially the removal of CO2 impurity, continues to grow because of the high-purity C2H2 required for various industrial applications. The adsorption separation of C2H2 and CO2 via porous materials is gaining a considerable attention as
Konstantinos E Papathanasiou et al.
Scientific reports, 7(1), 4743-4743 (2017-07-08)
Easy-to-prepare drug delivery systems, based on smart, silica gels have been synthesized, characterized, and studied as hosts in the controlled release of bisphosphonates. They exhibit variable release rates and final % release, depending on the nature of bisphosphonate (side-chain length
A S Malkani et al.
Science advances, 6(45) (2020-11-08)
Electrolyte cations affect the activity of surface-mediated electrocatalytic reactions; however, understanding the modes of interaction between cations and reaction intermediates remains lacking. We show that larger alkali metal cations (excluding the thickness of the hydration shell) promote the electrochemical CO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service