Skip to Content
Merck
All Photos(1)

Key Documents

232068

Sigma-Aldrich

Cesium hydroxide solution

50 wt. % in H2O, 99% trace metals basis

Synonym(s):

Cesium hydroxide hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$164.00
500 G
$560.00

$164.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$164.00
500 G
$560.00

About This Item

Linear Formula:
CsOH
CAS Number:
Molecular Weight:
149.91
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
liquid

$164.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

99% trace metals basis

form

liquid

concentration

50 wt. % in H2O

impurities

≤15000.0 ppm Trace Metal Analysis

density

1.72 g/mL at 25 °C

SMILES string

[OH-].[Cs+]

InChI

1S/Cs.H2O/h;1H2/q+1;/p-1

InChI key

HUCVOHYBFXVBRW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cesium hydroxide solution can be used to adjust the pH of rinsesolutions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brenda Farrell et al.
The Journal of general physiology, 152(7) (2020-05-10)
Outer hair cell (OHC) electromotility amplifies acoustic vibrations throughout the frequency range of hearing. Electromotility requires that the lateral membrane protein prestin undergo a conformational change upon changes in the membrane potential to produce an associated displacement charge. The magnitude
M M V Ramana et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 152, 165-171 (2015-07-25)
In the present work, isopropyl-6-amino-4-(3,5-bis(trifluoromethyl)phenyl)-5-cyano-2-methyl-4H-pyran-3-carboxylate (4H-pyran analog) has been synthesized by a three component reaction catalyzed by CsOH/γ-Al2O3 and characterized. The interaction of 4H-pyran analog with herring sperm DNA (hs DNA) under physiological conditions (phosphate buffer of pH 7.2) was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service