Skip to Content
Merck
All Photos(1)

Key Documents

400858

Sigma-Aldrich

Iron(II) iodide

AnhydroBeads, −10 mesh, ≥99.99% trace metals basis

Synonym(s):

Ferrous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeI2
CAS Number:
Molecular Weight:
309.65
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
synthesis grade
form:
beads
solubility:
H2O: soluble(lit.)
diethyl ether: soluble(lit.)
ethanol: soluble(lit.)

grade

for synthesis
synthesis grade

product line

AnhydroBeads

Assay

≥99.99% trace metals basis

form

beads

impurities

≤100.0 ppm Trace Metal Analysis

particle size

−10 mesh

solubility

H2O: soluble(lit.)
diethyl ether: soluble(lit.)
ethanol: soluble(lit.)

density

5.32 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

I[Fe]I

InChI

1S/Fe.2HI/h;2*1H/q+2;;/p-2

InChI key

BQZGVMWPHXIKEQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service