Skip to Content
Merck
All Photos(2)

Key Documents

208493

Sigma-Aldrich

Aluminum iodide

technical grade, 95%

Synonym(s):

Aluminum triiodide, Aluminum(III) iodide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$188.00

$188.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$188.00

About This Item

Linear Formula:
AlI3
CAS Number:
Molecular Weight:
407.70
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
technical grade
form:
chunks

$188.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

vapor pressure

1 mmHg ( 178 °C)

Assay

95%

form

chunks

bp

360 °C (lit.)

mp

191 °C (lit.)

density

3.98 g/mL at 25 °C (lit.)

SMILES string

I[Al](I)I

InChI

1S/Al.3HI/h;3*1H/q+3;;;/p-3

InChI key

CECABOMBVQNBEC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

The compound may be used as an additive for electrolytes in lithium ion batteries. [1][2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium air battery : alternate energy resource for the future.
Zahoor A, et al.
Journal of Electrochemical Science and Technology, 3(1), 14-23 (2012)
Cheap and environmentally benign electrochemical energy storage and conversion devices based on AlI3 electrolytes.
Xue B, et al.
Journal of the American Chemical Society, 128(27), 8720-8721 (2006)
Dien Li et al.
Journal of environmental radioactivity, 222, 106372-106372 (2020-08-11)
There are few effective technologies for the sequestration of highly water-soluble pertechnetate (TcO4-) from contaminated water despite the urgency of environmental and public health concerns. In this work, anion exchanged and cetyltrimethylammonium bromide (CTAB) functionalized MIL-101-Cr-NO3 were investigated for perrhenate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service