Skip to Content
Merck
All Photos(1)

Key Documents

399299

Sigma-Aldrich

trans-4-Methoxy-β-nitrostyrene

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$135.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$135.00

About This Item

Linear Formula:
CH3OC6H4CH=CHNO2
CAS Number:
Molecular Weight:
179.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

86-88 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, yellow

functional group

amine
nitro

SMILES string

[H]\C(=C(\[H])[N+]([O-])=O)c1ccc(OC)cc1

InChI

1S/C9H9NO3/c1-13-9-4-2-8(3-5-9)6-7-10(11)12/h2-7H,1H3/b7-6+

InChI key

JKQUXSHVQGBODD-VOTSOKGWSA-N

General description

trans-4-Methoxy-β-nitrostyrene participates in the Michael reaction on carbapenam intermediate.[1]

Application

trans-4-Methoxy-β-nitrostyrene acts as guest molecule and forms guest molecules forming co-crystal phases with the d-form of robust syndiotactic polystyrene (sPS).[2] It may be employed as a Michael acceptor in the synthesis of proline based chiral ionic liquid catalysts with two five-membered unsaturated aza-heterocycles.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer co-crystalline films for photonics.
Daniel C, et al.
Journal of the European Optical Society: Rapid Publications, 4 (2009)
Proline Based Chiral Ionic Liquids for Enantioselective Michael Reaction.
Nobuoka K, et al.
Organic Chemistry International (2014)
Sachin A Pawar et al.
Organic & biomolecular chemistry, 11(48), 8294-8297 (2013-11-13)
Herein, we report the development of mild, organocatalyzed routes to novel carbapenam derivatives through aldol, Mannich and Michael C-C bond forming reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service