Skip to Content
Merck
All Photos(1)

Key Documents

39915

Sigma-Aldrich

Heptakis(2,6-di-O-methyl)-β-cyclodextrin

≥98.0% (TLC)

Synonym(s):

2,6-Di-O-methyl-β-cyclodextrin, Dimethyl β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H98O35
CAS Number:
Molecular Weight:
1331.36
Beilstein:
1679144
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

optical activity

[α]20/D +158±3°, c = 10% in H2O

impurities

≤3% water

SMILES string

COC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](OC)[C@H](O[C@@H]3COC)O[C@H]4[C@H](O)[C@@H](OC)[C@H](O[C@@H]4COC)O[C@H]5[C@H](O)[C@@H](OC)[C@H](O[C@@H]5COC)O[C@H]6[C@H](O)[C@@H](OC)[C@H](O[C@@H]6COC)O[C@H]7[C@H](O)[C@@H](OC)[C@H](O[C@@H]7COC)O[C@H]8[C@H](O)[C@@H](OC)[C@H](O[C@@H]8COC)O[C@H]1[C@H](O)[C@H]2OC

InChI

1S/C56H98O35/c1-64-15-22-36-29(57)43(71-8)50(78-22)86-37-23(16-65-2)80-52(45(73-10)30(37)58)88-39-25(18-67-4)82-54(47(75-12)32(39)60)90-41-27(20-69-6)84-56(49(77-14)34(41)62)91-42-28(21-70-7)83-55(48(76-13)35(42)63)89-40-26(19-68-5)81-53(46(74-11)33(40)61)87-38-24(17-66-3)79-51(85-36)44(72-9)31(38)59/h22-63H,15-21H2,1-14H3/t22-,23-,24-,25-,26-,27-,28-,29+,30+,31+,32+,33+,34+,35+,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,56-/m1/s1

InChI key

QGKBSGBYSPTPKJ-UZMKXNTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a water-soluble derivative of cyclodextrin that exhibits the ability to effectively dissolve lipophilic drugs like steroid hormones, vitamins A, E, and K, etc. It can be used to improve the solubility and bioavailability of weakly hydrophilic pharmaceutical compounds.[1]

Application

Heptakis(2,6-di-O-methyl)-β-cyclodextrin may be used:
  • To measure the equilibrium constant of its complexes with various enantiomeric pairs of chiral nitroxides by electron paramagnetic resonance (EPR) spectroscopy.[2]
  • To study the crystal structure of its complex with n-butyl acrylate and isobornyl acrylate using X-ray diffraction (XRD).[3]
  • As a chiral selector to resolve amphetamine related drugs[4] and reduced haloperidol[5] by capillary zone electrophoresis.
  • As a component of thermosensitive polyurethane copolymers for their subsequent analysis by gel permeation chromatography (GPC).[6]

Reactant involved in:
  • Physicochemical studies of inclusion complexes
  • Probing diffusion and single molecule interactions with reconstituted membrane proteins
  • Interactions with micelles causing micellar rupture
  • Studying cellular transport, used as a cell penetration enhancer
  • Physicochemical and biopharmaceutical improvement of drugs particularly with solubilization and stability
  • Interactions with cholesterol

Other Notes

Complexes are more soluble in aqueous solution than the corresponding complexes of β-cyclodextrin[7]; Stimulation of fatty acid synthesis[8]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Casu et al.
Carbohydrate Research, 76, 59-59 (1979)
Effects of cyclodextrins on fatty acid synthesis.
Y Machida et al.
The Journal of biological chemistry, 248(17), 6246-6247 (1973-09-10)
New thermogelling copolymers composed of heptakis (2, 6-di-O-methyl)-beta-cyclodextrin, poly (propylene glycol), and poly (ethylene glycol)
Yang C, et al.
Journal of Materials Chemistry, 19(22), 3755-3763 (2009)
Chiral separation of reduced haloperidol by capillary zone electrophoresis with heptakis (2, 6-di-O-methyl)-beta-cyclodextrin
Wu HL, et al.
Journal of Liquid Chromatography and Related Technologies, 19(10), 1567-1577 (1996)
X-ray diffraction analysis of butyl-and isobornyl acrylate/heptakis (2, 6-di-O-methyl)-beta-cyclodextrin complexes and correlation to 1H NMR-spectra
Glockner P, et al.
Designed Monomers and Polymers, 5(2-3), 163-172 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service