Skip to Content
Merck
All Photos(2)

Key Documents

39832

Sigma-Aldrich

3,3-Dimethyl-1-butene

≥98.5% (GC)

Synonym(s):

Neohexene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$808.00

$808.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$808.00

About This Item

Linear Formula:
(CH3)3CCH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1697101
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$808.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

vapor density

>1 (vs air)

vapor pressure

6.96 psi ( 20 °C)

Assay

≥98.5% (GC)

refractive index

n20/D 1.376 (lit.)
n20/D 1.377

bp

41 °C (lit.)

mp

−115 °C (lit.)

density

0.653 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C=C

InChI

1S/C6H12/c1-5-6(2,3)4/h5H,1H2,2-4H3

InChI key

PKXHXOTZMFCXSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,3-Dimethyl-1-butene oxidizes selectively to corresponding epoxide by chlorine promoted Ag(111) oxygen adatoms.[1] 3,3-Dimethyl-1-butene undergoes copolymerization with ethylene using a chain-walking Pd-diimine catalyst.[2] Cationic polymerization of 3,3-dimethyl-1-butene (tert-butyl ethylene) has been carried out using AlCl3catalyst in ethyl chloride solvent in the temperature range -31 to -130°C.[3]

Application

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cationic isomerization polymerization of 3, 3-dimethyl-1-butene.
Kennedy JP, et al.
Makromol. Chem., 79(1), 109-121 (1964)
Peng Xiang et al.
Macromolecular rapid communications, 31(12), 1083-1089 (2011-05-19)
In this Communication, the copolymerization of ethylene with a sterically hindered α-olefin comonomer, γ-trisubstituted 3,3-dimethyl-1-butene (DMB), using a chain-walking Pd-diimine catalyst, [(ArNC(Me)(Me)CNAr)Pd(CH(3) )(NCMe)]SbF(6) (Ar2,6-(iPr)(2) C(6) H(3) ) (1) is reported. In spite of its high steric bulkiness in the close
Molecular mechanism of alkene epoxidation: A model study with 3, 3-dimethyl-1-butene on Ag (111).
Mukoid C, et al.
Catalysis Letters, 4(1), 57-61 (1990)

Articles

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service