Skip to Content
Merck
All Photos(3)

Key Documents

230545

Sigma-Aldrich

1-Hexene

97%

Synonym(s):

1-n-Hexene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$64.90
1 L
$77.90

$64.90


Estimated to ship onMarch 17, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$64.90
1 L
$77.90

About This Item

Linear Formula:
CH3(CH2)3CH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1209240
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$64.90


Estimated to ship onMarch 17, 2025


Request a Bulk Order

vapor density

3 (vs air)

Quality Level

vapor pressure

155 mmHg ( 21.1 °C)

Assay

97%

form

liquid

autoignition temp.

487 °F

refractive index

n20/D 1.388 (lit.)

bp

60-66 °C (lit.)

density

0.678 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

CCCCC=C

InChI

1S/C6H12/c1-3-5-6-4-2/h3H,1,4-6H2,2H3

InChI key

LIKMAJRDDDTEIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Hexene is primarily used in the synthesis of poly(1-hexene) and a variety of polyethylene copolymers.[1][2][3]

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-Catalyzed Enantioselective C-3 Allylation of 3-Substituted-1 H-Indoles Using Trialkylboranes.
Trost, Barry M and Quancard, Jean
Journal of the American Chemical Society, 128(19), 6314-6315 (2006)
Stereospecific living Ziegler- Natta polymerization of 1-hexene.
Jayaratne, Kumudini C and Sita, Lawrence R
Journal of the American Chemical Society, 122(5), 958-959 (2000)
Isospecific living polymerization of 1-hexene by a readily available nonmetallocene C 2-symmetrical zirconium catalyst.
Tshuva, Edit Y et al.
Journal of the American Chemical Society, 122(43), 10706-10707 (2000)
Chia-Hsiu Chen et al.
Journal of the American Chemical Society, 137(21), 6965-6971 (2015-05-12)
The stereochemistry, kinetics, and mechanism of olefin polymerization catalyzed by a set of zirconium-based metallocenes was studied by NMR using dissolution dynamic nuclear polarization (DNP). Hyperpolarized 1-hexene was polymerized in situ with a C2 symmetric catalyst, [(EBI)ZrMe][B(C6F5)4] (EBI = rac-(C2H4(1-indenyl)2))
Yuichi Ito et al.
Implant dentistry, 24(4), 477-479 (2015-06-04)
The evaluation of bone quality at the site of the alveolar bone for a dental implant is very important. This study presents an easy technique for direct evaluation of alveolar bone quality using nondecalcified cryofilm frozen sections on human alveolar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service