Skip to Content
Merck
All Photos(1)

Documents

394483

Sigma-Aldrich

Cyclobutyl methyl ketone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7COCH3
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

137-139 °C/754 mmHg (lit.)

density

0.902 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C1CCC1

InChI

1S/C6H10O/c1-5(7)6-3-2-4-6/h6H,2-4H2,1H3

InChI key

JPJOOTWNILDNAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclobutyl methyl ketone (CMK, methyl cyclobutyl ketone) is an alicyclic ketone. It has been synthesized by reacting cyclobutanecarbonyl chloride with magnesium salt of malonic ester. The mechanism of the photochemical reaction of gaseous mixtures of cyclobutyl methyl ketone diluted in argon has been studied by steady-state and pulsed (laser) photolysis. It′s infrared (3500 to 30cm-1) and Raman (3200 to 30cm-1) spectra in gaseous and solid state has been recorded. The UV spectra have been measured in alcohol and isooctane solution. Study on the vapour phase thermal and photodecomposition of methyl cyclobutyl ketone shows that the mode of decomposition is free radical in nature.

Application

Cyclobutyl methyl ketone may be used as a model compound to study the oxidation mechanism of pinonic acid by hydroxyl radicals.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Thermal Decomposition of Methyl Cyclobutyl Ketone1,2.
Daignault LG and Walters WD
Journal of the American Chemical Society, 80(3), 541-545 (1958)
The photochemistry of cyclobutyl methyl ketone. Part 1.-Room-temperature results and the general mechanism.
Baldwin PJ, et al
J. Chem. Soc., Faraday II, 83(6), 1049-1058 (1987)
Azo-bis Nitriles. 1 Decomposition of Azo Compounds. A Special Case of Carbon-Carbon Hyperconjugation in a Free Radical Reaction.
Overberger CG and Lebovits A
Journal of the American Chemical Society, 76(10), 2722-2725 (1954)
Structure and Reactivity in the Vapor Phase Photolysis of Ketones. III. Methyl Cyclobutyl Ketone1,2.
Norman I and Pitts Jr. JN
Journal of the American Chemical Society, 77(23), 6104-6107 (1955)
Spectra and structure of small ring molecules: Part XLIV. Vibrational spectra and conformational stability of cyclobutylmethyl ketone.
Durig JR, et al
Journal of Molecular Structure, 125(3-4), 211-227 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service