Skip to Content
Merck
All Photos(1)

Key Documents

389951

Sigma-Aldrich

Pentyl isocyanate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$203.00

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$203.00

About This Item

Linear Formula:
CH3(CH2)4NCO
CAS Number:
Molecular Weight:
113.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

136-137 °C (lit.)

density

0.878 g/mL at 25 °C (lit.)

functional group

amine
isocyanate

SMILES string

CCCCCN=C=O

InChI

1S/C6H11NO/c1-2-3-4-5-7-6-8/h2-5H2,1H3

InChI key

VRVUKQWNRPNACD-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lajos Nagy et al.
International journal of molecular sciences, 21(4) (2020-02-23)
In this article, the capability of encoding information using a homologous series of monodisperse monomethoxypolyethylene glycols (mPEG), with a number of ethylene oxide units ranging from nEO = 5 to 8, and monodisperse linear aliphatic isocyanates containing a number of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service