Skip to Content
Merck
All Photos(1)

Key Documents

282790

Sigma-Aldrich

Benzo-15-crown-5

98%

Synonym(s):

2,3,5,6,8,9,11,12-Octahydro-1,4,7,10,13-benzopentaoxacyclopentadecin, 2,3-Benzo-1,4,7,10,13-pentaoxacyclopentadecane, Benzo15C5, Monobenzo-15-crown-5

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$203.00

$203.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$203.00

About This Item

Empirical Formula (Hill Notation):
C14H20O5
CAS Number:
Molecular Weight:
268.31
Beilstein:
1624106
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

$203.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Quality Level

Assay

98%

mp

78-80 °C (lit.)

functional group

ether

SMILES string

C1COCCOc2ccccc2OCCOCCO1

InChI

1S/C14H20O5/c1-2-4-14-13(3-1)18-11-9-16-7-5-15-6-8-17-10-12-19-14/h1-4H,5-12H2

InChI key

FNEPSTUXZLEUCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzo-15-crown-5 is a macrocyclic compound that belongs to the class of crown ethers. It is used as a ligand in metal-catalyzed reactions to enhance the reactivity and selectivity by coordinating with metal cations. For instance, it has been utilized in certain metal-catalyzed cross-coupling reactions, such as Suzuki-Miyaura or Stille reactions, to improve the efficiency and selectivity of the transformations. Benzo-15-crown is also employed as a phase-transfer catalyst in organic synthesis.

Application

Benzo-15-crown-5 can be used as a reactant in the synthesis of a phenol-based benzo-15-crown-5 ether resin. The resulting resin further used for lithium isotope separation, which relies on the selective absorption of the Li-6 isotope.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Patrick Shupp et al.
Dalton transactions (Cambridge, England : 2003), 46(28), 9163-9171 (2017-07-05)
We report the synthesis, interconversions and X-ray structures of a set of [mFe-nS]-type carbonyl clusters (where S = S
Li-Jing Du et al.
Food chemistry, 262, 118-128 (2018-05-13)
In this study, dispersive micro solid phase extraction (DMSPE) combined with ultrahigh-performance liquid chromatography coupled with quadrupole time-of-flight tandem mass spectrometry method was established to extract and determine sudan dyes and their metabolites in real samples. The crown ether microfunctionalized
Shinzo Kagabu et al.
Bioorganic & medicinal chemistry letters, 19(11), 2947-2948 (2009-05-05)
Imidacloprid (IMI) derivatives conjugated with benzo-15-crown-5 and benzo-18-crown-6 structures, applied for the first time to explore novel insecticidal molecule, elicited strong excitatory toxic signs to the house flies and stunningly exhibited three to five times higher insecticidal activity than that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service