Skip to Content
Merck
All Photos(1)

Key Documents

384275

Sigma-Aldrich

2,3,5-Trichloropyridine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$196.00

$196.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$196.00

About This Item

Empirical Formula (Hill Notation):
C5H2Cl3N
CAS Number:
Molecular Weight:
182.44
Beilstein:
119384
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$196.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

bp

219 °C (lit.)

mp

46-50 °C (lit.)

functional group

chloro

SMILES string

Clc1cnc(Cl)c(Cl)c1

InChI

1S/C5H2Cl3N/c6-3-1-4(7)5(8)9-2-3/h1-2H

InChI key

CNLIIAKAAMFCJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Separation and migration behavior of 2,3,5-trichloropyridine has been studied by micellar electrokinetic chromatography.[1] Crystal structure of 2,3,5-trichloropyridine has been reported. Mol­ecules of 2,3,5-trichloropyridine in crystal are stacked along the short a axis and forms a layer structure.[1] 2,3,5-Trichloropyridine is reported to undergo nucleophilic displacement reaction in ionic liquid to afford the corresponding 2-aryloxylpropionate.[2]

Application

2,3,5-Trichloropyridine may be used in the synthesis of 3,5-dichloro-2-arylpyridines via palladium acetate-catalyzed ligand-free Suzuki reaction with arylboronic acids.[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Formation of 2, 3, 5-Trichloropyridine and its Nucleophilic Displacement Reactions in Ionic Liquid.
Zhong P, et al.
Synthetic Communications, 34(23), 4301-4311 (2004)
Huanan Hu et al.
Molecules (Basel, Switzerland), 14(9), 3153-3160 (2009-09-29)
A highly efficient palladium acetate-catalyzed ligand-free Suzuki reaction of 2,3,5-trichloropyridine with arylboronic acids in aqueous phase was developed. High yields of 3,5-dichloro-2-arylpyridines, a simple Pd source, absence of ligands, and environmentally benign as well as mild reaction conditions are important
C E Lin et al.
Journal of chromatography. A, 910(1), 165-171 (2001-03-27)
The separation and migration behavior of pyridine and eight chloropyridines, including three monochloropyridines, four dichloropyridines, and 2,3,5-trichloropyridine were investigated by micellar electrokinetic chromatography using either sodium dodecyl sulfate (SDS) as an anionic surfactant or SDS-Brij 35 mixed micelles. Various parameters
2, 3, 5-Trichloropyridine.
Ma H-F, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(1), o311-o312 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service