Skip to Content
Merck
All Photos(2)

Key Documents

D73707

Sigma-Aldrich

2,6-Dichloropyridine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$86.00

$86.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$86.00

About This Item

Empirical Formula (Hill Notation):
C5H3Cl2N
CAS Number:
Molecular Weight:
147.99
Beilstein:
108664
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

98%

form

solid

mp

83-86 °C (lit.)

SMILES string

Clc1cccc(Cl)n1

InChI

1S/C5H3Cl2N/c6-4-2-1-3-5(7)8-4/h1-3H

InChI key

FILKGCRCWDMBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service