Skip to Content
Merck
All Photos(1)

Key Documents

380903

Sigma-Aldrich

7-Methylbenzo[a]pyrene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14
CAS Number:
Molecular Weight:
266.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

219-220 °C (lit.)

SMILES string

Cc1cccc2c1cc3ccc4cccc5ccc2c3c45

InChI

1S/C21H14/c1-13-4-2-7-17-18-11-10-15-6-3-5-14-8-9-16(12-19(13)17)21(18)20(14)15/h2-12H,1H3

InChI key

PYVWGNPFWVQISD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidized metabolites of 7-methylbenzo[a]pyrene.
M Konieczny et al.
Carcinogenesis, 3(5), 573-575 (1982-01-01)
P L Chiu et al.
Chemico-biological interactions, 52(3), 265-277 (1985-01-01)
The enantiomers of trans-7,8-dihydroxy-7,8-dihydro-7-methylbenzo[a]pyrene (7-MBaP 7,8-dihydrodiol) and of trans-7,8-dihydroxy-7,8,9,10-tetrahydro-7-methylbenzo[a]pyrene (7-MBaP 7,8-tetrahydrodiol) were directly resolved by high-performance liquid chromatography (HPLC) using a commercially available column packed with an (R)-N-(3,5-dinitrobenzoyl)-phenylglycine derivative of gamma-aminopropylsilanized silica. The absolute configurations of the resolved enantiomers were
T Kinoshita et al.
Cancer research, 42(10), 4032-4038 (1982-10-01)
The ultimate carcinogenic form of benzo(a)pyrene (BP) is thought to result from metabolic activation at the 7 to 10 positions. Substitution by a methyl group at these positions would be expected to inhibit strongly their metabolism even though 7-methylbenzo(a)pyrene (7-MeBP)
William B Studabaker et al.
Journal of chromatography. A, 1492, 106-116 (2017-03-08)
Development of the Athabasca Oil Sands Region in northeastern Alberta, Canada has contributed polycyclic aromatic hydrocarbons (PAHs) and polycyclic aromatic compounds (PACs), which include alkyl PAHs and dibenzothiophenes, to the regional environment. A new analytical method was developed for quantification
T K Wong et al.
Chemico-biological interactions, 36(2), 153-166 (1981-08-01)
The 7-methylbenzo[a]pyrene (7-MBaP) was incubated with liver microsomes of rats pretreated with polychlorinated biphenyls (Aroclor 1254) (PCBs). Metabolites of 7-MBaP were isolated by both reversed-phase and normal-phage high performance liquid chromatography (HPLC) and were characterized by nuclear magnetic resonance, UV-visible

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service