Skip to Content
Merck
All Photos(1)

Key Documents

37275

Sigma-Aldrich

(+)-Dihydrocarvone

mixture of isomers

Synonym(s):

(2R,5R)-5-Isopropenyl-2-methylcyclohexanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$80.60

$80.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$80.60

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2044615
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

$80.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

optical activity

[α]20/D +20±2°, neat

composition

n-(+)-dihydrocarvone, ~77%
iso-(+)-dihydrocarvone, ~20%

refractive index

n20/D 1.471

density

0.928 g/mL at 20 °C (lit.)

functional group

ketone

SMILES string

CC1CCC(CC1=O)C(C)=C

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h8-9H,1,4-6H2,2-3H3

InChI key

AZOCECCLWFDTAP-UHFFFAOYSA-N

General description

(+)-Dihydrocarvone, a monoterpenoid compound[1] found in caraway oil,[2] is a key building block to synthesize sesquiterpenes.[3] It is generally produced either by the hydrogenation of carvone or oxidation of limonene.[2]

Application

(+)-Dihydrocarvone may be used in the following processes:
  • Synthesis of dispiro 1,2,4,5-tetraoxanes, which show potent anti-malarial activity.[4]
  • Synthesis of an epoxylactone by oxidation, which can undergo copolymerization with ε-caprolactone to form cross-linked copolymers with shape memory properties.[2]
  • Synthesis of α-Cyperone, a eudesmane type sesquiterpenoid compound with potent insecticidal activity.[5]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The structure and antimalarial activity of dispiro-1, 2, 4, 5-tetraoxanes derived from (+)-dihydrocarvone.
Dong Y, et al.
Bioorganic & Medicinal Chemistry Letters, 20(22), 6359-6361 (2010)
Oxidized dihydrocarvone as a renewable multifunctional monomer for the synthesis of shape memory polyesters.
Lowe JR, et al.
Biomacromolecules, 10(7), 2003-2008 (2009)
Insecticidal activity of sesquiterpenes skeleton synthesized by the conventional Robinson annulations reaction on Drosophila melanogaster.
Alarcon J, et al.
Industrial Crops and Products, 42, 268-272 (2013)
Dan Jin et al.
Scientific reports, 10(1), 3309-3309 (2020-02-26)
Cannabis research has historically focused on the most prevalent cannabinoids. However, extracts with a broad spectrum of secondary metabolites may have increased efficacy and decreased adverse effects compared to cannabinoids in isolation. Cannabis's complexity contributes to the length and breadth
Redife Aslihan Ucar et al.
Food microbiology, 91, 103454-103454 (2020-06-17)
This research determined the concentration of trehalose, xylose and l-citrulline in fresh and fermented cucumbers and their utilization by Lactobacillus pentosus, Lactobacillus plantarum, Lactobacillus brevis and Lactobacillus buchneri. Targeted compounds were measured by HPLC and the ability of the lactobacilli

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service