Skip to Content
Merck
All Photos(1)

Key Documents

W296236

Sigma-Aldrich

(−)-Isopulegol

≥98%

Synonym(s):

(1R,2S,5R)-2-Isopropenyl-5-methylcyclohexanol, (1R,3R,4S)-p-Menth-8-en-3-ol

Sign Into View Organizational & Contract Pricing

Select a Size

W296236-SAMPLE
$54.60
1 KG
$161.00
9 KG
$1,200.00
20 KG
$2,510.00

$54.60


Estimated to ship onMay 28, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W296236-SAMPLE
$54.60
1 KG
$161.00
9 KG
$1,200.00
20 KG
$2,510.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2962
Beilstein:
1906573
EC Number:
Council of Europe no.:
2033
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.067
NACRES:
NA.21
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

$54.60


Estimated to ship onMay 28, 2025


Request a Bulk OrderRequest more information

biological source

synthetic

grade

Halal

Assay

≥98%

optical activity

[α]20/D −21°, neat

refractive index

n20/D 1.471 (lit.)

bp

212 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

minty; cooling

SMILES string

C[C@@H]1CC[C@H]([C@H](O)C1)C(C)=C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h8-11H,1,4-6H2,2-3H3/t8-,9+,10-/m1/s1

InChI key

ZYTMANIQRDEHIO-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Isopulegol is a monoterpenic alcohol widely used in flavor and fragrance industry. It is also a key intermediate in the synthesis of the industrially important chemical, (−)-menthol.[1]

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclization of (+)-citronellal to (?)-isopulegol catalyzed by H 3 PW 12 O 40/SiO 2.
da Silva KA, et al.
Catalysis Communications, 5(8), 425-429 (2004)
David Olagnier et al.
Bioorganic & medicinal chemistry letters, 17(22), 6075-6078 (2007-10-02)
Pure natural monoterpenes were evaluated in vitro for their antiplasmodial activities against Plasmodium falciparum. Chemically modified terpenes were also tested to see whether the introduction of an alkyne, a cyclopropane, a diene, or a cyclopentenone moiety had an influence on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service