Skip to Content
Merck
All Photos(1)

Key Documents

372293

Sigma-Aldrich

Benzo-18-crown-6

98%

Synonym(s):

18-Benzo-6-crown ether, 2,3,5,6,8,9,11,12,14,15-Decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecin, 2,3-Benzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2-ene, Benzo18C6, Monobenzo-18-crown-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O6
CAS Number:
Molecular Weight:
312.36
Beilstein:
1626123
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

42-45 °C (lit.)

SMILES string

C1COCCOCCOc2ccccc2OCCOCCO1

InChI

1S/C16H24O6/c1-2-4-16-15(3-1)21-13-11-19-9-7-17-5-6-18-8-10-20-12-14-22-16/h1-4H,5-14H2

InChI key

DSFHXKRFDFROER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elsayed M Zahran et al.
The Analyst, 142(17), 3241-3249 (2017-08-11)
Electrospray ionization mass spectrometry ESI-MS is a powerful technique for the characterization of macromolecules and their noncovalent binding with guest ions. We herein evaluate the feasibility of using ESI-MS as a screening tool for predicting potentiometric selectivities of ionophores. Ion-selective
Shinzo Kagabu et al.
Bioorganic & medicinal chemistry letters, 19(11), 2947-2948 (2009-05-05)
Imidacloprid (IMI) derivatives conjugated with benzo-15-crown-5 and benzo-18-crown-6 structures, applied for the first time to explore novel insecticidal molecule, elicited strong excitatory toxic signs to the house flies and stunningly exhibited three to five times higher insecticidal activity than that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service