Skip to Content
Merck
All Photos(1)

Key Documents

371459

Sigma-Aldrich

1H,1H,2H-Perfluoro-1-hexene

99%

Synonym(s):

3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$114.00

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$114.00

About This Item

Linear Formula:
CH2=CH(CF2)3CF3
CAS Number:
Molecular Weight:
246.07
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.22

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

8.5 (vs air)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.3 (lit.)

bp

59-60 °C (lit.)

density

1.452 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C

InChI

1S/C6H3F9/c1-2-3(7,8)4(9,10)5(11,12)6(13,14)15/h2H,1H2

InChI key

GVEUEBXMTMZVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Angevine Malley et al.
Inhalation toxicology, 14(8), 773-787 (2002-07-18)
Inhalation studies were conducted to determine the potential subchronic toxicity of a mixture of trans-1,2-dichloroethylene (70%), cis-1,2-dichloroethylene (5%), and perfluorobutylethylene (25%). Groups of rats were exposed to 0, 400, 2000, or 8000 ppm concentrations of the mixture vapor 6 h/day
T Nakayama et al.
The journal of physical chemistry. A, 111(5), 909-915 (2007-02-03)
FTIR-smog chamber techniques were used to study the products of the Cl atom and OH radical initiated oxidation of CF3CH=CH2 in 700 Torr of N2/O2, diluent at 296 K. The Cl atom initiated oxidation of CF3CH=CH2 in 700 Torr of
G L Kennedy
Critical reviews in toxicology, 21(2), 149-170 (1990-01-01)
Fluorine-containing monomers form the basis for production of a large number of commercially important polymers. Most of the polymerization occurs as gas-phase reactions, hence the hazards associated with the monomers arises primarily from inhalation. The chemicals covered in this review

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service