Skip to Content
Merck
All Photos(1)

Key Documents

370614

Sigma-Aldrich

4-(Trifluoromethoxy)benzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CH2OH
CAS Number:
Molecular Weight:
192.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

108 °C/25 mmHg (lit.)

density

1.326 g/mL at 25 °C (lit.)

functional group

fluoro
hydroxyl

SMILES string

OCc1ccc(OC(F)(F)F)cc1

InChI

1S/C8H7F3O2/c9-8(10,11)13-7-3-1-6(5-12)2-4-7/h1-4,12H,5H2

InChI key

ZLSOZAOCYJDPKX-UHFFFAOYSA-N

General description

4-(Trifluoromethoxy)benzyl alcohol is an organic building block.

Application

4-(Trifluoromethoxy)benzyl alcohol may be used in chemical synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service