Skip to Content
Merck
All Photos(1)

Key Documents

369330

Sigma-Aldrich

N-tert-Butyltrimethylsilylamine

98%

Synonym(s):

N-(Trimethylsilyl)-tert-butylamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$191.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$191.00

About This Item

Linear Formula:
(CH3)3SiNHC(CH3)3
CAS Number:
Molecular Weight:
145.32
Beilstein:
1734528
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.406 (lit.)

bp

118-119 °C (lit.)

density

0.762 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(C)(C)N[Si](C)(C)C

InChI

1S/C7H19NSi/c1-7(2,3)8-9(4,5)6/h8H,1-6H3

InChI key

GVHIREZHTRULPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

N-tert-Butyltrimethylsilylamine (Me3SiNH-t-Bu) reacts with [Cp*Rh(H2O)3](OTf)2 in acetone to yield a hydroxyl-capped half-cubane [Cp*3Rh3(μ-OH)3(μ3-OH)](OTf)3(t-BuNH3).[1]

Application

N-tert-Butyltrimethylsilylamine may be used in the preparation of selenium and tellurium chalcogenides[2] and the facile synthesis of N-alkylaminomethylphosphonates.[3]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Won Seok Han et al.
Dalton transactions (Cambridge, England : 2003), (20)(20), 3360-3364 (2004-10-16)
Treatment of [Cp*Rh(H(2)O)(3)](OTf)(2) (1) with Me(3)SiNH-t-Bu in acetone gave a hydroxyl-capped half-cubane [Cp*(3)Rh(3)(mu-OH)(3)(mu(3)-OH)](OTf)(3)(t-BuNH(3)) (2). Slow diffusion of Me(3)SiN(3) in diethyl ether into compound in acetone produced an azido-capped half-cubane [Cp*(3)Rh(3)(mu-N(3))(3)(mu(3)-N(3))](OTf)(2) (3). On the other hand, treating 1 with Me(3)SiN(3) in
J. Solid State Inorg. Chem., 29, 759-759 (1992)
Synthetic Communications, 21, 201-201 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service