Skip to Content
Merck
All Photos(2)

Key Documents

368512

Sigma-Aldrich

7-Hydroxy-4-(trifluoromethyl)coumarin

98%

Synonym(s):

4-(Trifluoromethyl)umbelliferone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5F3O3
CAS Number:
Molecular Weight:
230.14
Beilstein:
210932
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

pKa 

7.26

mp

178-180 °C (lit.)

fluorescence

λex 385 nm; λem 502 nm in methanol

SMILES string

Oc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C10H5F3O3/c11-10(12,13)7-4-9(15)16-8-3-5(14)1-2-6(7)8/h1-4,14H

InChI key

CCKWMCUOHJAVOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Hydroxy-4-(trifluoromethyl)coumarin is a class of 7-hydroxycoumarin that is majorly used as laser dyes. It has a characterized emission spectra and may be used in excited state proton transfer (ESPT).[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean Ekins et al.
Drug metabolism and disposition: the biological fate of chemicals, 31(9), 1077-1080 (2003-08-16)
CYP2D6 and CYP3A4 represent two particularly important members of the cytochrome p450 enzyme family due to their involvement in the metabolism of many commercially available drugs. Avoiding potent inhibitory interactions with both of these enzymes is highly desirable in early
Base-induced phototautomerization in 7-hydroxy-4-(trifluoromethyl) coumarin
Westlake BC, et al.
The Journal of Physical Chemistry B, 116(51), 14886-14891 (2012)
Hideto Jinno et al.
Drug metabolism and disposition: the biological fate of chemicals, 31(5), 528-532 (2003-04-16)
UDP-glucuronosyltransferase (UGT) 1A10 is an isoform of UGT1A, which is expressed in extrahepatic, biliary and aerodigestive/gastrointestinal tissues. We have previously reported two nonsynonymous single nucleotide polymorphisms in exon 1 of human UGT1A10 gene; 177G>A and 605C>T resulting in amino acid
Risto O Juvonen et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(8), 885-893 (2020-01-07)
Sulfonation is an important high affinity elimination pathway for phenolic compounds.In this study sulfonation of 7-hydroxycoumarin and 13 its derivatives were evaluated in liver cytosols of human and six animal species. 7-hydroxycoumarin and its derivatives are strongly fluorescent, and their
Nobumitsu Hanioka et al.
Life sciences, 87(7-8), 261-268 (2010-07-14)
UDP-glucuronosyltransferase 1A1 (UGT1A1) plays important roles in the glucuronidation of various drugs and endogenous substances. Cynomolgus monkeys are regarded as experimental animals closer to humans in studies on safety evaluation and biotransformation for drug development. In this study, the similarities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service