Skip to Content
Merck
All Photos(1)

Key Documents

89803

Supelco

Methoxyamine hydrochloride

for GC derivatization, LiChropur, 97.5-102.5% (AT)

Synonym(s):

O-Methylhydroxylamine hydrochloride, Methoxylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$58.50
5 G
$225.00

$58.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$58.50
5 G
$225.00

About This Item

Linear Formula:
CH3ONH2 · HCl
CAS Number:
Molecular Weight:
83.52
Beilstein:
3589723
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.05

$58.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

97.5-102.5% (AT)

form

crystals

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

mp

151-154 °C (lit.)

SMILES string

Cl.CON

InChI

1S/CH5NO.ClH/c1-3-2;/h2H2,1H3;1H

InChI key

XNXVOSBNFZWHBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoxyamine hydrochloride is a commonly used reagent for the generation of oxime derivatives of aldehyde- or ketone-group containing compounds. It is typically employed as an oximation reagent prior to silylation derivatizations.[1][2]

Application

Methoxyamine hydrochloride can be used as:
  • A reagent for the preparation of O-methyl oximes.
  • A reagent used to make O-methyl oximes from aldehydes or ketones.
  • An alkylation reagent.
  • It may be used as a derivatization agent to improve the sensitivity of opioids,[3][4] steroids[5] and ionic metabolites[6] in biological samples and non-volatile flavor compounds in "Angeleno" plum fruit[7] prior to the determination of analytes using gas chromatography-mass spectrometry (GC-MS) technique.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted derivatization: application to steroid profiling by gas chromatography/mass spectrometry
Casals G, et al.
Journal of Chromatography. B, Biomedical Applications, 960(8), 8-13 (2014)
GC-MS analysis of multiply derivatized opioids in urine
Chen BG, et al.
Journal of Mass Spectrometry : Jms, 42(8), 1012-1023 (2007)
Solid-phase analytical derivatization for gas-chromatography-mass-spectrometry-based metabolomics
Takeo E, et al.
Journal of Bioscience and Bioengineering, 124(6), 700-706 (2017)
Data correction strategy for metabolomics analysis using gas chromatography-mass spectrometry.
Kanani HH and Klapa MI
Metabolic engineering, 9(1), 39-51 (2007)
Optimization of Precolumn Derivatization for Simultaneous Gas Chromatography-Mass Spectrometric Analysis of Non-volatile Flavor Compounds in" Angeleno" Plum Fruit [J]
HU L, et al.
Food Science, 14(8), 8-13 (2010)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service