Skip to Content
Merck
All Photos(1)

Key Documents

366536

Sigma-Aldrich

Ammonium nitrate-15N

98 atom % 15N

Synonym(s):

15N ammonium salt, 15N labeled ammonium nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$500.00

$500.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$500.00

About This Item

Linear Formula:
NH415NO3
CAS Number:
Molecular Weight:
81.04
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

$500.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 15N

form

solid

bp

210 °C (lit.)

mp

169 °C (lit.)

mass shift

M+1

SMILES string

O[N+]([O-])=O.[H][N+]([H])([H])[H]

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3/i1+1;

InChI key

PRORZGWHZXZQMV-YTBWXGASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium nitrate-15N is an inorganic salt containing 15N isotope

Application

Ammonium nitrate-15N is a commonly used compound in agriculture research as a tracer.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurence V Bindschedler et al.
Phytochemistry, 69(10), 1962-1972 (2008-06-10)
Hydroponic isotope labelling of entire plants (HILEP) is a cost-effective method enabling metabolic labelling of whole and mature plants with a stable isotope such as (15)N. By utilising hydroponic media that contain (15)N inorganic salts as the sole nitrogen source
Sally Taylor et al.
Pest management science (2018-04-01)
Current insect resistance management plans rely on refuges of plants without Bacillus thuringiensis (Bt) toxins to provide a gene pool of unexposed insects. Insects from refuges must mate with insects from Bt maize to slow resistance evolution. We used stable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service