Skip to Content
Merck
All Photos(4)

Key Documents

316830

Sigma-Aldrich

Urea-15N2

98 atom % 15N, 99% (CP)

Synonym(s):

15N Labeled Urea, 15N Labeled urea, Carbamide-15N2, Carbonyldiamide-15N2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$364.00
1 G
$1,250.00

$364.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$364.00
1 G
$1,250.00

About This Item

Linear Formula:
H215NCO15NH2
CAS Number:
Molecular Weight:
62.04
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.12

$364.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

mp

132-135 °C (lit.)

application(s)

agriculture

mass shift

M+2

storage temp.

room temp

SMILES string

[15NH2]C([15NH2])=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i2+1,3+1

InChI key

XSQUKJJJFZCRTK-SUEIGJEOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Urea-15N2 enriched with 15N isotope.

Application

Urea-15N2 is used
  • In agriculture research as a technique to determine uptake or denitrification and leaching losses of native soil N via the “added N interaction".
  • In horticulture, massive labeling of trees to ascertain that N originating from 15N leaf labeling.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa Scholefield et al.
Frontiers in molecular neuroscience, 14, 711396-711396 (2021-11-10)
Widespread elevations in brain urea have, in recent years, been reported in certain types of age-related dementia, notably Alzheimer's disease (AD) and Huntington's disease (HD). Urea increases in these diseases are substantive, and approximate in magnitude to levels present in
Xu Zhao et al.
Journal of agricultural and food chemistry, 64(15), 3017-3024 (2016-03-30)
The nitrogen (N) isotope method reveals that application of fertilizer N can increase crop uptake or denitrification and leaching losses of native soil N via the "added N interaction". However, there is currently little evidence of the impact of added
Sungchul Kim et al.
Nature, 579(7797), 141-145 (2020-02-23)
CRISPR-Cas immunity protects prokaryotes against invading genetic elements1. It uses the highly conserved Cas1-Cas2 complex to establish inheritable memory (spacers)2-5. How Cas1-Cas2 acquires spacers from foreign DNA fragments (prespacers) and integrates them into the CRISPR locus in the correct orientation

Related Content

Tracer techniques using isotopic N have been employed since the 1940s to investigate such topics as N transformations and cycling in agricultural and other soils, N sources for crop uptake, the fate and behavior of fertilizer N applied to soils, the nature and extent of soil and fertilizer losses, and fertilizer N uptake efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service