Skip to Content
Merck
All Photos(1)

Key Documents

363006

Sigma-Aldrich

Ammonium-15N acetate

98 atom % 15N

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$749.00

$749.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$749.00

About This Item

Linear Formula:
CH3CO215NH4
CAS Number:
Molecular Weight:
78.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$749.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 15N

Quality Level

form

solid

mp

110-112 °C (dec.) (lit.)

mass shift

M+1

SMILES string

CC([O-])=O.[H][15N+]([H])([H])[H]

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3/i;1+1

InChI key

USFZMSVCRYTOJT-IEOVAKBOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Journal of the American Chemical Society, 107, 3706-3706 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service