Skip to Content
Merck
All Photos(1)

Key Documents

362794

Sigma-Aldrich

Cholesterol

≥99%

Synonym(s):

3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Beilstein:
1915888
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥99%

optical activity

[α]20/D −40°, c = 2 in chloroform

bp

360 °C (lit.)

mp

147-149 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinjiang Fan et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(23), 7261-7266 (2015-06-04)
Translocator protein (TSPO) is a key member of the mitochondrial cholesterol transport complex in steroidogenic tissues. To assess the function of TSPO, we generated two lines of Cre-mediated Tspo conditional knockout (cKO) mice. First, gonadal somatic cell-targeting Amhr2-Cre mice were
Steven A Grover et al.
The lancet. Diabetes & endocrinology, 3(2), 114-122 (2014-12-09)
Despite the increased risk of cardiovascular disease and type 2 diabetes associated with excess bodyweight, development of a clinically meaningful metric for health professionals remains a challenge. We estimated the years of life lost and the life-years lost from diabetes
Bo Angelin et al.
Journal of internal medicine, 277(3), 331-342 (2014-04-24)
Liver-selective thyromimetic agents could provide a new approach for treating dyslipidaemia. We performed a multicentre, randomized, placebo-controlled, double-blind study to evaluate the efficacy and safety of eprotirome, a liver-selective thyroid hormone receptor agonist, in 98 patients with primary hypercholesterolaemia. After
Michael J Pencina et al.
The New England journal of medicine, 370(15), 1422-1431 (2014-03-22)
The 2013 guidelines of the American College of Cardiology and the American Heart Association (ACC-AHA) for the treatment of cholesterol expand the indications for statin therapy for the prevention of cardiovascular disease. Using data from the National Health and Nutrition
Matthew P Buman et al.
American journal of epidemiology, 179(3), 323-334 (2013-12-10)
Sleep and sedentary and active behaviors are linked to cardiovascular disease risk biomarkers, and across a 24-hour day, increasing time in 1 behavior requires decreasing time in another. We explored associations of reallocating time to sleep, sedentary behavior, or active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service